FRO091856475 Frontier Scientific Services Inc.

N-Ethyl-N-Methyl-2-(Piperidin-4-Yl-Oxy)-Acetamide

  • MDL number MFCD08061073
  • Molecular Weight : 200.2831

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart