FRO091856480 Frontier Scientific Services Inc.

2-Piperazin-1-Yl-N-Propyl-Nicotinamide

  • MDL number MFCD08064264
  • Molecular Weight : 248.3306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart