FRO091856485 Frontier Scientific Services Inc.

2-Chloro-4-(Piperidin-3-Yl-Oxy)-Benzonitrile

  • MDL number MFCD08061078
  • Molecular Weight : 236.7032

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart