FRO091856508 Frontier Scientific Services Inc.

4-[2-(Propan-2-Yl)Phenoxy]Piperidine

  • MDL number MFCD06247929
  • Molecular Weight : 219.322601

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart