FRO091856515 Frontier Scientific Services Inc.

2-[4-(3-Fluorophenoxymethyl)Phenyl]Acetic Acid

  • MDL number MFCD08061043
  • Molecular Weight : 260.260315

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart