FRO091856521 Frontier Scientific Services Inc.

2-Chloro-N-(1-Phenyl-Ethyl)-Acetamide

  • MDL number MFCD00040691
  • Cas Number : 13230-80-3
  • Molecular Weight : 197.66624

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart