FRO091856534 Frontier Scientific Services Inc.

N,N-Dimethyl-2-(Piperidin-4-Yl-Oxy)-Benzamide

  • MDL number MFCD08061086
  • Molecular Weight : 248.3277

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart