FRO091856535 Frontier Scientific Services Inc.

4-(3-Chloro-Phenoxy)-Piperidine

  • MDL number MFCD08061087
  • Cas Number : 97840-40-9
  • Molecular Weight : 211.688004

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart