FRO091856546 Frontier Scientific Services Inc.

3-Methoxy-4-(Piperidin-4-Yl-Oxy)-Benzonitrile

  • MDL number MFCD08061089
  • Molecular Weight : 232.2847

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart