FRO091856555 Frontier Scientific Services Inc.

2-Methyl-1-(4-Methylamino-Piperidin-1-Yl)-Propan-1-One

  • MDL number MFCD08061054
  • Molecular Weight : 184.2837

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart