FRO091856587 Frontier Scientific Services Inc.

3-(3-Methoxyphenoxy)Piperidine

  • MDL number MFCD08060989
  • Cas Number : 902837-25-6
  • Molecular Weight : 207.268906

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart