FRO091856588 Frontier Scientific Services Inc.

Propionic Acid Sodium Salt

  • MDL number MFCD00002759
  • Cas Number : 137-40-6
  • Molecular Weight : 96.0621

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Sodium Salt
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart