FRO091856589 Frontier Scientific Services Inc.

Ammonium Thiocyanate Ammonium Salt

  • MDL number MFCD00011428
  • Cas Number : 1762-95-4
  • Molecular Weight : 76.1198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Ammonium Salt
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart