FRO091856594 Frontier Scientific Services Inc.

1-(4-Methoxy-Phenyl)-2-Methylamino-Ethanol

  • MDL number MFCD08060969
  • Molecular Weight : 181.2366

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart