FRO091856618 Frontier Scientific Services Inc.

3-(2-Ethoxyphenoxy)Piperidine

  • MDL number MFCD08061107
  • Molecular Weight : 221.295502

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart