FRO091856704 Frontier Scientific Services Inc.

Methyl 2,2-Dimethyl-4,6-Dioxocyclohexane-1-Carboxylate

  • MDL number MFCD00154564
  • Cas Number : 59373-32-9
  • Molecular Weight : 198.215805

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart