FRO091856721 Frontier Scientific Services Inc.

1-Chloro-5-Methoxy-2-Pentene�

  • MDL number MFCD00018970
  • Cas Number : 3577-82-0
  • Molecular Weight : 134.607

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart