FRO091856722 Frontier Scientific Services Inc.

2-Chlorophenylhydrazine-4-Sulfonic Acid�

  • MDL number MFCD00025091
  • Cas Number : 6628-76-8
  • Molecular Weight : 222.6513

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart