FRO091856729 Frontier Scientific Services Inc.

1-(2-Ethylcrotonoyl)Urea�

  • MDL number MFCD00025443
  • Cas Number : 5982-97-8
  • Molecular Weight : 156.1859

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart