FRO091856732 Frontier Scientific Services Inc.

2-(1-Hydroxyethyl)Cyclohexanol

  • MDL number MFCD00019359
  • Cas Number : 60713-86-2
  • Molecular Weight : 144.2155

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart