FRO091856800 Frontier Scientific Services Inc.

N-(O-Tolyl)-Diacetoacetamide

  • MDL number MFCD00026265
  • Cas Number : 68912-00-5
  • Molecular Weight : 275.307

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart