FRO091856807 Frontier Scientific Services Inc.

2,3-Diphenylpropanenitrile

  • MDL number MFCD00034964
  • Cas Number : 3333-14-0
  • Molecular Weight : 207.270401

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart