FRO091856875 Frontier Scientific Services Inc.

(E)-3-(2,3-DIMETHOXY-PHENYL)-2-METHYL-ACRYLIC ACID

  • MDL number MFCD00666042
  • Molecular Weight : 222.243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart