FRO091856894 Frontier Scientific Services Inc.

1,1-Bis(4-Chlorophenyl)-1,2,2,2-Tetrachloroethane

  • MDL number MFCD00018831
  • Cas Number : 3563-45-9
  • Molecular Weight : 388.9379

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart