FRO091857082 Frontier Scientific Services Inc.

1-[(2-Fluorophenyl)Carbonyl]Piperazine

  • MDL number MFCD01821526
  • Cas Number : 1011-15-0
  • Molecular Weight : 208.232101

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart