FRO091857096 Frontier Scientific Services Inc.

(2R)-1-(Benzyl-Oxy)Butan-2-Amine

  • MDL number MFCD00800550
  • Cas Number : 142559-11-3
  • Molecular Weight : 179.258804

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart