FRO152357906 Frontier Scientific Services Inc.

M-TOLYL ISOTHIOCYANATE

  • MDL number MFCD00004807
  • Molecular Weight : 149.21569

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart