FRO152357920 Frontier Scientific Services Inc.

4-(Ethoxymethyl)Piperidine

  • MDL number MFCD08274968
  • Cas Number : 265108-38-1
  • Molecular Weight : 143.2267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart