FRO152358011 Frontier Scientific Services Inc.

6-Bromo-2-Methyl-Quinoline-4-Thiol

  • MDL number MFCD01143227
  • Molecular Weight : 254.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart