FRO152358046 Frontier Scientific Services Inc.

3-Chloro-6-(2-Methly-Benzyl)-Pyridazine

  • MDL number MFCD00214803
  • Molecular Weight : 218.6879

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart