FRO152358085 Frontier Scientific Services Inc.

2-Butylphenylhydrazine Hydrochloride

  • MDL number MFCD08459595
  • Molecular Weight : 200.7124

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart