FRO152358268 Frontier Scientific Services Inc.

3-Chloro-6-(Pyridin-2-Yl-Methyl)Pyridazine

  • MDL number MFCD00140708
  • Cas Number : 78784-70-0
  • Molecular Weight : 205.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart