FRO152358277 Frontier Scientific Services Inc.

4-(3,4-Dichlorophenoxy)Piperidine Hydrochloride

  • MDL number MFCD03840126
  • Cas Number : 817186-93-9
  • Molecular Weight : 282.593996

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart