FRO152358298 Frontier Scientific Services Inc.

N-Propyl-Guanidine Hemi-Sulfate

  • MDL number MFCD09839059
  • Molecular Weight : 150.191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hemi-Sulfate
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart