FRO152358304 Frontier Scientific Services Inc.

3-Amino-N-Methyl-3-Thioxopropanamide

  • MDL number MFCD00778360
  • Cas Number : 59749-86-9
  • Molecular Weight : 132.1852

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart