FRO152358326 Frontier Scientific Services Inc.

5-Methylpyrazine-2-Carboxamidine

  • MDL number MFCD04116535
  • Molecular Weight : 136.1575

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart