FRO152358334 Frontier Scientific Services Inc.

Cycloheptanecarboxamidine

  • MDL number MFCD05662921
  • Molecular Weight : 140.2301

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart