FRO152358346 Frontier Scientific Services Inc.

2-(1-Methyl-1H-Benzimidazol-2-Yl)Ethanethioamide

  • MDL number MFCD05722969
  • Cas Number : 61689-99-4
  • Molecular Weight : 205.2833

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart