FRO152358417 Frontier Scientific Services Inc.

2-Fluorophenethylisocyanide

  • MDL number MFCD04117532
  • Molecular Weight : 149.16921

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart