FRO152358425 Frontier Scientific Services Inc.

1-(3-Isocyanopropyl)-1H-Imidazole

  • MDL number MFCD06200540
  • Molecular Weight : 135.16988

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart