FRO152358500 Frontier Scientific Services Inc.

Pht-Gly-Leu-Oh

  • MDL number MFCD00070130
  • Cas Number : 6707-71-7
  • Molecular Weight : 318.3323

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart