FRO152358518 Frontier Scientific Services Inc.

2-(Methylsulfanyl)Pyrido[4,3-D][1,3]Oxazole

  • MDL number MFCD08447111
  • Molecular Weight : 166.199997

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart