FRO154961997 Frontier Scientific Services Inc.

2-(Piperidin-4-Yl-Oxy)Acetamide

  • MDL number MFCD08061069
  • Cas Number : 912761-65-0
  • Molecular Weight : 158.198303

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart