FRO154962005 Frontier Scientific Services Inc.

4-Hydrazinyl-7-Methoxy-2-Methylquinoline

  • MDL number MFCD00272480
  • Molecular Weight : 203.240402

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart