FRO182845587 Frontier Scientific Services Inc.

3-(3-Cyano-Phenyl)-5-Ethyl-4,6-Dioxo-Octahydro-Pyrrolo[3,4-C]Pyrrole-1-Carboxylic Acid Methyl Ester

  • MDL number MFCD11656687
  • Molecular Weight : 327.3427

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart