FRO182845716 Frontier Scientific Services Inc.

3-Amino-3-(4-Fluorophenyl)Propanamide

  • MDL number MFCD06216203
  • Molecular Weight : 182.1992

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart