FRO182845725 Frontier Scientific Services Inc.

4-[3-Methyl-5-(2-Methylphenyl)-4H-1,2,4-Triazol-4-Yl]Phenol

  • MDL number MFCD11101069
  • Molecular Weight : 265.3175

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart