FRO182845776 Frontier Scientific Services Inc.

(2-Chloro(3-Pyridyl))-N-(5-Methylisoazol-3-Yl)Formamide

  • MDL number MFCD00245702
  • Molecular Weight : 237.6472

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart