FRO182845788 Frontier Scientific Services Inc.

1-(2-Chloro-6-Fluorophenyl)Cyclopentanecarboxylic Acid

  • MDL number MFCD00800630
  • Molecular Weight : 242.676

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart