FRO182845802 Frontier Scientific Services Inc.

[1-(6-Chloro-Pyridin-2-Yl)-1H-[1,2,4]Triazol-3-Yl]-Acetic Acid Ethyl Ester

  • MDL number MFCD00202370
  • Molecular Weight : 266.688892

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart